Assay Details
Gene Symbol: rassf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004550.1 | NP_001004550.1 | 3 - 4 | 571 | 73 | |
XM_005173951.4 | XP_005174008.1 | 4 - 5 | 1213 | 73 | ||
XM_005173952.4 | XP_005174009.1 | 2 - 3 | 473 | 73 | ||
XM_017353113.2 | XP_017208602.1 | 4 - 5 | 932 | 73 | ||
GenBank mRNA | BC081661.1 | - | 3 - 4 | 571 | 73 | |
BC093276.1 | - | 2 - 3 | 388 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
