Assay Details
| Gene Symbol: adprs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004565.2 | NP_001004565.2 | 3 - 4 | 845 | 109 | |
| GenBank mRNA | BC081683.1 | - | 4 - 5 | 772 | 109 | |
| BC152161.1 | - | 3 - 4 | 620 | 109 | ||
| BC164177.1 | - | 3 - 4 | 483 | 109 | ||
| EH450150.1 | - | 3 - 4 | 875 | 109 | ||
| EH475955.1 | - | 3 - 4 | 767 | 109 | ||
More Information
Set Membership: |
|

adprs
Genomic Map