Assay Details
| Gene Symbol: adprs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004565.2 | NP_001004565.2 | 4 - 5 | 1030 | 126 | |
| XM_009293135.3 | XP_009291410.1 | 2 - 3 | 330 | 126 | ||
| GenBank mRNA | BC081683.1 | - | 5 - 6 | 126 | ||
| BC152161.1 | - | 4 - 5 | 126 | |||
| BC164177.1 | - | 4 - 5 | 126 | |||
| EH475955.1 | - | 2 - 3 | 582 | 126 | ||
| EH612280.1 | - | 2 - 3 | 126 | |||
More Information
Set Membership: |
|

adprs
Genomic Map