Assay Details
| Gene Symbol: etfdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004598.1 | NP_001004598.1 | 12 - 13 | 1807 | 61 | |
| GenBank mRNA | BC081588.1 | - | 12 - 13 | 1807 | 61 | |
More Information
Set Membership: |
|

etfdh
Genomic Map