Assay Details
| Gene Symbol: rbm39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012304.1 | NP_001012304.1 | 10 - 11 | 1140 | 78 | |
| XM_005169275.3 | XP_005169332.1 | 11 - 12 | 1126 | 78 | ||
| XM_005169277.4 | XP_005169334.1 | 11 - 12 | 1217 | 78 | ||
| XM_005169278.4 | XP_005169335.1 | 12 - 13 | 1185 | 78 | ||
| XM_009306024.3 | XP_009304299.1 | 12 - 13 | 1416 | 78 | ||
| XM_021479585.1 | XP_021335260.1 | 11 - 12 | 1687 | 78 | ||
| GenBank mRNA | BC044487.1 | - | 10 - 11 | 1140 | 78 | |
| BC063981.1 | - | 9 - 10 | 1623 | 78 | ||
| BC097076.1 | - | 12 - 13 | 1363 | 78 | ||
| BC165689.1 | - | 9 - 10 | 909 | 78 | ||
More Information
Set Membership: |
|

rbm39a
Genomic Map