Assay Details
| Gene Symbol: rbm39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012304.1 | NP_001012304.1 | 15 - 16 | 1737 | 62 | |
| XM_005169275.3 | XP_005169332.1 | 16 - 17 | 1723 | 62 | ||
| XM_005169277.4 | XP_005169334.1 | 16 - 17 | 1814 | 62 | ||
| XM_005169278.4 | XP_005169335.1 | 17 - 18 | 1782 | 62 | ||
| XM_009306024.3 | XP_009304299.1 | 17 - 18 | 2013 | 62 | ||
| XM_021479585.1 | XP_021335260.1 | 16 - 17 | 2284 | 62 | ||
| GenBank mRNA | BC044487.1 | - | 15 - 16 | 1737 | 62 | |
| BC063981.1 | - | 14 - 15 | 2220 | 62 | ||
| BC097076.1 | - | 17 - 18 | 1960 | 62 | ||
| BC165689.1 | - | 14 - 15 | 1506 | 62 | ||
More Information
Set Membership: |
|

rbm39a
Genomic Map