Assay Details
| Gene Symbol: rbm39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012304.1 | NP_001012304.1 | 5 - 6 | 614 | 148 | |
| XM_005169275.3 | XP_005169332.1 | 6 - 7 | 600 | 148 | ||
| XM_005169277.4 | XP_005169334.1 | 6 - 7 | 691 | 148 | ||
| XM_005169278.4 | XP_005169335.1 | 7 - 8 | 659 | 148 | ||
| XM_009306024.3 | XP_009304299.1 | 8 - 9 | 809 | 148 | ||
| XM_021479585.1 | XP_021335260.1 | 6 - 7 | 1161 | 148 | ||
| GenBank mRNA | BC044487.1 | - | 5 - 6 | 614 | 148 | |
| BC097076.1 | - | 8 - 9 | 756 | 148 | ||
| BC165689.1 | - | 4 - 5 | 383 | 148 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009306024.3, XM_021479585.1 transcripts.
Set Membership: |
|

rbm39a
Genomic Map