Assay Details
| Gene Symbol: bhmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012480.1 | NP_001012498.1 | 7 - 8 | 1084 | 72 | |
| GenBank mRNA | AY830415.1 | - | 7 - 8 | 1084 | 72 | |
| BC109472.1 | - | 7 - 8 | 1095 | 72 | ||
More Information
Set Membership: |
|

bhmt
Genomic Map