Assay Details
| Gene Symbol: natd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013318.1 | NP_001013336.1 | 2 - 3 | 264 | 66 | |
| GenBank mRNA | BC090291.1 | - | 2 - 3 | 264 | 66 | |
More Information
Set Membership: |
|

natd1
Genomic Map