Assay Details
| Gene Symbol: psmd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017624.2 | NP_001017624.1 | 3 - 4 | 295 | 76 | |
| GenBank mRNA | BC092825.1 | - | 3 - 4 | 296 | 76 | |
| BC164468.1 | - | 3 - 4 | 316 | 76 | ||
More Information
Set Membership: |
|

psmd4b
Genomic Map