Assay Details
| Gene Symbol: ppp1r11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017627.1 | NP_001017627.1 | 1 - 2 | 441 | 109 | |
| GenBank mRNA | BC092823.1 | - | 1 - 2 | 441 | 109 | |
| BC164924.1 | - | 1 - 2 | 113 | 109 | ||
More Information
Set Membership: |
|

ppp1r11
Genomic Map