Assay Details
| Gene Symbol: prkn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017635.1 | NP_001017635.1 | 10 - 11 | 1172 | 122 | |
| GenBank mRNA | BC093281.1 | - | 10 - 11 | 1172 | 122 | |
| BC164537.1 | - | 9 - 10 | 1186 | 122 | ||
More Information
Set Membership: |
|

prkn
Genomic Map