Assay Details
| Gene Symbol: enpp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017887.1 | NP_001017887.1 | 2 - 3 | 1029 | 66 | |
| GenBank mRNA | BC093443.1 | - | 2 - 3 | 1029 | 66 | |
More Information
Set Membership: |
|

enpp4
Genomic Map