Assay Details
| Gene Symbol: cert1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017914.4 | NP_001017914.3 | 3 - 4 | 394 | 66 | |
| NM_001281872.1 | NP_001268801.1 | 3 - 4 | 394 | 66 | ||
| GenBank mRNA | EU000165.1 | - | 3 - 4 | 337 | 66 | |
| EU000166.1 | - | 3 - 4 | 337 | 66 | ||
More Information
Set Membership: |
|

cert1a
Genomic Map