Assay Details
| Gene Symbol: dpysl5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018149.1 | NP_001018159.1 | 11 - 12 | 1594 | 106 | |
| GenBank mRNA | AY987371.1 | - | 11 - 12 | 1594 | 106 | |
| BC124748.1 | - | 11 - 12 | 1545 | 106 | ||
| BC165313.1 | - | 10 - 11 | 1468 | 106 | ||
More Information
Set Membership: |
|

dpysl5a
Genomic Map