Assay Details
Gene Symbol: lrit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020536.1 | NP_001018372.2 | 2 - 3 | 929 | 80 | |
GenBank mRNA | BC095086.1 | - | 2 - 3 | 929 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
