Assay Details
| Gene Symbol: zgc:112294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020677.1 | NP_001018513.1 | 2 - 3 | 262 | 89 | |
| XM_017356090.2 | XP_017211579.1 | 2 - 3 | 327 | 89 | ||
| GenBank mRNA | BC095738.1 | - | 2 - 3 | 262 | 89 | |
More Information
Set Membership: |
|

zgc:112294
Genomic Map