Assay Details
Gene Symbol: bpnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037568.1 | NP_001032657.1 | 3 - 4 | 931 | 99 | |
GenBank mRNA | BC110106.1 | - | 3 - 4 | 931 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
