Assay Details
| Gene Symbol: ca14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328144.1 | NP_001315073.1 | 11 - 12 | 1174 | 67 | |
| NR_137291.1 | 10 - 11 | 1047 | 67 | |||
| GenBank mRNA | BC107639.1 | - | 10 - 11 | 1040 | 67 | |
More Information
Set Membership: |
|

ca14
Genomic Map