Assay Details
Gene Symbol: scn4aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039825.1 | NP_001034914.1 | 16 - 17 | 2883 | 69 | |
XM_017358479.2 | XP_017213968.1 | 17 - 18 | 3069 | 69 | ||
GenBank mRNA | DQ149506.1 | - | 16 - 17 | 2883 | 69 | |
DQ351532.1 | - | 5 - 6 | 1071 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
