Assay Details
Gene Symbol: scn4aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039825.1 | NP_001034914.1 | 18 - 19 | 3217 | 75 | |
XM_017358479.2 | XP_017213968.1 | 19 - 20 | 3403 | 75 | ||
GenBank mRNA | DQ149506.1 | - | 18 - 19 | 3214 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
