Assay Details
Gene Symbol: scn4aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039825.1 | NP_001034914.1 | 21 - 22 | 3677 | 126 | |
XM_017358479.2 | XP_017213968.1 | 22 - 23 | 3863 | 126 | ||
GenBank mRNA | DQ149506.1 | - | 21 - 22 | 3674 | 126 | |
DQ351532.1 | - | 10 - 11 | 1865 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
