Assay Details
| Gene Symbol: stim1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044799.1 | NP_001038264.1 | 5 - 6 | 756 | 113 | |
| XM_005173392.4 | XP_005173449.1 | 5 - 6 | 806 | 113 | ||
| XM_005173393.4 | XP_005173450.1 | 5 - 6 | 806 | 113 | ||
| XM_009291871.3 | XP_009290146.1 | 5 - 6 | 806 | 113 | ||
| GenBank mRNA | BC134856.1 | - | 5 - 6 | 718 | 113 | |
More Information
Set Membership: |
|

stim1a
Genomic Map