Assay Details
| Gene Symbol: stim1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044799.1 | NP_001038264.1 | 6 - 7 | 871 | 73 | |
| XM_005173392.4 | XP_005173449.1 | 6 - 7 | 921 | 73 | ||
| XM_005173393.4 | XP_005173450.1 | 6 - 7 | 921 | 73 | ||
| XM_009291871.3 | XP_009290146.1 | 6 - 7 | 921 | 73 | ||
More Information
Set Membership: |
|

stim1a
Genomic Map