Assay Details
| Gene Symbol: selenoo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044871.3 | NP_001038336.2 | 6 - 7 | 1897 | 76 | |
| GenBank mRNA | EE689105.1 | - | 2 - 3 | 233 | 76 | |
More Information
Set Membership: |
|

selenoo1
Genomic Map