Assay Details
Gene Symbol: pi4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045485.2 | NP_001038950.1 | 6 - 7 | 1196 | 64 | |
GenBank mRNA | AY929291.1 | - | 6 - 7 | 1033 | 64 | |
BC129195.1 | - | 6 - 7 | 1196 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
