Assay Details
| Gene Symbol: lsm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048006.1 | NP_001041471.1 | 3 - 4 | 184 | 67 | |
| GenBank mRNA | BC055642.1 | - | 2 - 3 | 187 | 67 | |
| BC133957.1 | - | 3 - 4 | 186 | 67 | ||
| BI846179.1 | - | 3 - 4 | 188 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC055642.1 transcripts.
Set Membership: |
|

lsm7
Genomic Map