Assay Details
Gene Symbol: abi1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076707.1 | NP_001070175.1 | 6 - 7 | 898 | 75 | |
XM_005171176.4 | XP_005171233.1 | 7 - 8 | 938 | 75 | ||
GenBank mRNA | BC124378.1 | - | 6 - 7 | 898 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
