Assay Details
Gene Symbol: rgs18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077307.1 | NP_001070775.1 | 5 - 6 | 755 | 88 | |
XM_005163237.4 | XP_005163294.1 | 4 - 5 | 514 | 88 | ||
XM_021481328.1 | XP_021337003.1 | 3 - 4 | 337 | 88 | ||
GenBank mRNA | BC124764.1 | - | 5 - 6 | 755 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
