Assay Details
Gene Symbol: usp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077449.1 | NP_001070917.1 | 39 - 40 | 6742 | 82 | |
XM_005167736.4 | XP_005167793.1 | 40 - 41 | 7242 | 82 | ||
XM_009304755.3 | XP_009303030.1 | 40 - 41 | 7243 | 82 | ||
XM_009304756.3 | XP_009303031.1 | 40 - 41 | 7243 | 82 | ||
XM_009304757.3 | XP_009303032.1 | 40 - 41 | 7244 | 82 | ||
GenBank mRNA | DQ086492.1 | - | 39 - 40 | 6742 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
