Assay Details
| Gene Symbol: usp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077449.1 | NP_001070917.1 | 44 - 45 | 7513 | 73 | |
| XM_009304755.3 | XP_009303030.1 | 46 - 47 | 8122 | 73 | ||
| XM_009304757.3 | XP_009303032.1 | 45 - 46 | 8015 | 73 | ||
| GenBank mRNA | DQ086492.1 | - | 44 - 45 | 7513 | 73 | |
More Information
Set Membership: |
|

usp9
Genomic Map