Assay Details
Gene Symbol: kdm6al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080193.1 | NP_001073662.1 | 16 - 17 | 2023 | 108 | |
XM_005168796.4 | XP_005168853.1 | 17 - 18 | 4859 | 108 | ||
GenBank mRNA | BC066564.1 | - | 17 - 18 | 2477 | 108 | |
BC129198.1 | - | 16 - 17 | 2023 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
