Assay Details
| Gene Symbol: arl6ip6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089501.1 | NP_001082970.1 | 3 - 4 | 666 | 95 | |
| GenBank mRNA | BC134166.1 | - | 3 - 4 | 666 | 95 | |
More Information
Set Membership: |
|

arl6ip6
Genomic Map