Assay Details
| Gene Symbol: rgs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199274.1 | NP_954968.1 | 3 - 4 | 299 | 85 | |
| GenBank mRNA | AY423029.1 | - | 3 - 4 | 299 | 85 | |
| BC092675.1 | - | 3 - 4 | 313 | 85 | ||
| BC141786.1 | - | 3 - 4 | 312 | 85 | ||
| BC154780.1 | - | 3 - 4 | 310 | 85 | ||
More Information
Set Membership: |
|

rgs4
Genomic Map