Assay Details
| Gene Symbol: rgs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199274.1 | NP_954968.1 | 4 - 5 | 466 | 60 | |
| GenBank mRNA | AY423029.1 | - | 4 - 5 | 466 | 60 | |
| BC141786.1 | - | 4 - 5 | 479 | 60 | ||
| BC154780.1 | - | 4 - 5 | 477 | 60 | ||
More Information
Set Membership: |
|

rgs4
Genomic Map