Assay Details
| Gene Symbol: antxr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102568.1 | NP_001096038.1 | 6 - 7 | 542 | 71 | |
| GenBank mRNA | EF591979.1 | - | 6 - 7 | 542 | 71 | |
More Information
Set Membership: |
|

antxr2b
Genomic Map