Assay Details
Gene Symbol: tlcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110382.1 | NP_001103852.1 | 3 - 4 | 707 | 135 | |
GenBank mRNA | BC154828.1 | - | 3 - 4 | 707 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
