Assay Details
Gene Symbol: mapk8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110389.1 | NP_001103859.1 | 5 - 6 | 653 | 150 | |
XM_005156857.4 | XP_005156914.1 | 6 - 7 | 876 | 150 | ||
GenBank mRNA | BC154588.1 | - | 5 - 6 | 653 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
