Assay Details
| Gene Symbol: sema3h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328378.1 | NP_001315307.1 | 9 - 10 | 1050 | 89 | |
| NM_131050.2 | NP_571125.2 | 9 - 10 | 1050 | 89 | ||
| XM_009303983.3 | XP_009302258.1 | 9 - 10 | 1405 | 89 | ||
| GenBank mRNA | AY860954.1 | - | 9 - 10 | 972 | 89 | |
| BC162689.1 | - | 9 - 10 | 1050 | 89 | ||
| CU638804.1 | - | 9 - 10 | 972 | 89 | ||
More Information
Set Membership: |
|

sema3h
Genomic Map