Assay Details
| Gene Symbol: hoxa11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131147.1 | NP_571222.1 | 1 - 2 | 614 | 84 | |
| GenBank mRNA | BC162917.1 | - | 1 - 2 | 638 | 84 | |
| BC162927.1 | - | 1 - 2 | 638 | 84 | ||
More Information
Set Membership: |
|

hoxa11b
Genomic Map