Assay Details
Gene Symbol: pax2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131184.2 | NP_571259.1 | 2 - 3 | 704 | 144 | |
XM_005156735.4 | XP_005156792.1 | 2 - 3 | 1925 | 144 | ||
XM_005156744.4 | XP_005156801.1 | 2 - 3 | 1347 | 144 | ||
XM_005156747.4 | XP_005156804.1 | 2 - 3 | 1340 | 144 | ||
XM_005156748.4 | XP_005156805.1 | 2 - 3 | 1341 | 144 | ||
XM_017358683.2 | XP_017214172.1 | 2 - 3 | 1922 | 144 | ||
XM_017358684.2 | XP_017214173.1 | 2 - 3 | 1437 | 144 | ||
XM_017358685.2 | XP_017214174.1 | 2 - 3 | 1358 | 144 | ||
XM_021480494.1 | XP_021336169.1 | 2 - 3 | 1937 | 144 | ||
XM_021480498.1 | XP_021336173.1 | 2 - 3 | 1934 | 144 | ||
XM_021480500.1 | XP_021336175.1 | 2 - 3 | 1452 | 144 | ||
XM_021480501.1 | XP_021336176.1 | 2 - 3 | 1372 | 144 | ||
XM_021480505.1 | XP_021336180.1 | 2 - 3 | 1360 | 144 | ||
XM_021480507.1 | XP_021336182.1 | 2 - 3 | 1353 | 144 | ||
XM_021480508.1 | XP_021336183.1 | 2 - 3 | 1354 | 144 | ||
XM_021480509.1 | XP_021336184.1 | 2 - 3 | 1347 | 144 | ||
XM_021480512.1 | XP_021336187.1 | 2 - 3 | 1353 | 144 | ||
XM_021480513.1 | XP_021336188.1 | 2 - 3 | 1340 | 144 | ||
GenBank mRNA | AF067530.1 | - | 2 - 3 | 200 | 144 | |
AF067531.1 | - | 2 - 3 | 200 | 144 | ||
AF067532.1 | - | 2 - 3 | 212 | 144 | ||
AF067533.1 | - | 2 - 3 | 200 | 144 | ||
AF067534.1 | - | 2 - 3 | 212 | 144 | ||
AF067535.1 | - | 2 - 3 | 200 | 144 | ||
AF067536.1 | - | 2 - 3 | 200 | 144 | ||
AF067537.1 | - | 2 - 3 | 212 | 144 | ||
AF067538.1 | - | 2 - 3 | 212 | 144 | ||
AF067539.1 | - | 2 - 3 | 200 | 144 | ||
AF067540.1 | - | 2 - 3 | 200 | 144 | ||
AF067541.1 | - | 2 - 3 | 212 | 144 | ||
BC066690.1 | - | 2 - 3 | 704 | 144 | ||
X63961.1 | - | 826 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
