Assay Details
| Gene Symbol: myod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131262.2 | NP_571337.2 | 2 - 3 | 773 | 96 | |
| GenBank mRNA | BC114261.1 | - | 2 - 3 | 773 | 96 | |
| Z36945.1 | - | 781 | 96 | |||
More Information
Set Membership: |
|

myod1
Genomic Map