Assay Details
| Gene Symbol: thrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131340.1 | NP_571415.1 | 7 - 8 | 526 | 93 | |
| XM_005159501.4 | XP_005159558.1 | 2 - 3 | 440 | 93 | ||
| XM_005159502.4 | XP_005159559.1 | 2 - 3 | 640 | 93 | ||
| XM_005159505.4 | XP_005159562.1 | 2 - 3 | 659 | 93 | ||
| XM_021467984.1 | XP_021323659.1 | 2 - 3 | 638 | 93 | ||
| XM_021467985.1 | XP_021323660.1 | 7 - 8 | 543 | 93 | ||
| XM_021467986.1 | XP_021323661.1 | 7 - 8 | 538 | 93 | ||
| XM_021467987.1 | XP_021323662.1 | 6 - 7 | 498 | 93 | ||
| XM_021467988.1 | XP_021323663.1 | 6 - 7 | 480 | 93 | ||
| XM_021467989.1 | XP_021323664.1 | 6 - 7 | 493 | 93 | ||
| XM_021467990.1 | XP_021323665.1 | 5 - 6 | 435 | 93 | ||
| XM_021467991.1 | XP_021323666.1 | 2 - 3 | 660 | 93 | ||
| GenBank mRNA | AB759513.1 | - | 2 - 3 | 440 | 93 | |
| AF109732.1 | - | 7 - 8 | 526 | 93 | ||
| AF322218.1 | - | 2 - 3 | 440 | 93 | ||
| BC163106.1 | - | 6 - 7 | 356 | 93 | ||
| BC163114.1 | - | 6 - 7 | 356 | 93 | ||
More Information
Set Membership: |
|

thrb
Genomic Map