Assay Details
Gene Symbol: ephb4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131414.1 | NP_571489.1 | 10 - 11 | 1861 | 94 | |
XM_021476001.1 | XP_021331676.1 | 10 - 11 | 2626 | 94 | ||
GenBank mRNA | AJ005026.2 | - | 10 - 11 | 1861 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
