Assay Details
| Gene Symbol: spon1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131517.1 | NP_571592.1 | 11 - 12 | 1658 | 87 | |
| XM_005174346.4 | XP_005174403.1 | 11 - 12 | 1954 | 87 | ||
| GenBank mRNA | AB006087.1 | - | 11 - 12 | 1658 | 87 | |
| BC162653.1 | - | 11 - 12 | 1512 | 87 | ||
| KM655691.1 | - | 11 - 12 | 1479 | 87 | ||
More Information
Set Membership: |
|

spon1b
Genomic Map