Assay Details
Gene Symbol: spon1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131517.1 | NP_571592.1 | 14 - 15 | 2167 | 64 | |
XM_005174346.4 | XP_005174403.1 | 14 - 15 | 2463 | 64 | ||
GenBank mRNA | AB006087.1 | - | 14 - 15 | 2167 | 64 | |
BC162653.1 | - | 14 - 15 | 2021 | 64 | ||
KM655691.1 | - | 14 - 15 | 1988 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
