Assay Details
| Gene Symbol: hoxa1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131536.2 | NP_571611.1 | 1 - 2 | 737 | 80 | |
| XM_017352263.2 | XP_017207752.1 | 2 - 3 | 2955 | 80 | ||
| GenBank mRNA | AJ306430.1 | - | 1 - 2 | 641 | 80 | |
| AJ306431.1 | - | 2 - 3 | 536 | 80 | ||
| BC162701.1 | - | 1 - 2 | 737 | 80 | ||
| DQ060531.1 | - | 1 - 2 | 236 | 80 | ||
More Information
Set Membership: |
|

hoxa1a
Genomic Map