Assay Details
| Gene Symbol: dedd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131602.2 | NP_571677.2 | 3 - 4 | 829 | 109 | |
| XM_017351359.1 | XP_017206848.1 | 3 - 4 | 831 | 109 | ||
| XM_021466747.1 | XP_021322422.1 | 3 - 4 | 825 | 109 | ||
| GenBank mRNA | AF232226.1 | - | 3 - 4 | 727 | 109 | |
| BC075865.1 | - | 3 - 4 | 726 | 109 | ||
More Information
Set Membership: |
|

dedd1
Genomic Map