Assay Details
| Gene Symbol: dedd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131602.2 | NP_571677.2 | 5 - 6 | 1071 | 77 | |
| XM_017351359.1 | XP_017206848.1 | 5 - 6 | 1073 | 77 | ||
| GenBank mRNA | AF232226.1 | - | 5 - 6 | 969 | 77 | |
More Information
Set Membership: |
|

dedd1
Genomic Map