Assay Details
| Gene Symbol: dedd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131602.2 | NP_571677.2 | 6 - 7 | 1456 | 93 | |
| XM_017351359.1 | XP_017206848.1 | 6 - 7 | 1458 | 93 | ||
| GenBank mRNA | AF232226.1 | - | 6 - 7 | 1354 | 93 | |
More Information
Set Membership: |
|

dedd1
Genomic Map